missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ ProDoma Hybridoma Media
Brand: Lonza™ BE02-029Q
This item is not returnable.
View return policy
Description
The original hybridoma media for murine, human, and chimeric origin cells.
- Non-animal, protein-free media for your hybridoma needs.
- Scalable from small flasks to large-scale bioreactors.
- Hybridoma cell growth
- Monoclonal antibody production

Specifications
2° to 8°C | |
ProDoma 1 | |
No Glutamine, No Phenol Red | |
1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ BioWhittaker™ ProDoma Hybridoma Media > ProDoma 1, 1L
Spot an opportunity for improvement?Share a Content Correction