missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension. BioWhittaker™ Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
Brand: Lonza™ BE17-160E
This item is not returnable.
View return policy
Description
- Clear, colorless-yellow
- Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
- Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
- Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
- Porcine parvovirus tested and mycoplasma screened
- Product is manufactured with irradiated trypsin
Specifications
-10° to -20°C | |
Sterile | |
6.44 to 8.04 |
Cell Culture Dissociation Reagent | |
No Calcium, No Magnesium | |
100 mL |