missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension. BioWhittaker™ Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
Specifications
Content And Storage | -10° to -20°C |
---|---|
Product Type | Cell Culture Dissociation Reagent |
Sterility | Sterile |
Description
- Clear, colorless-yellow
- Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
- Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
- Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
- Porcine parvovirus tested and mycoplasma screened
- Product is manufactured with irradiated trypsin
Specifications
-10° to -20°C | |
Cell Culture Dissociation Reagent | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ BioWhittaker™ Trypsin Reagent