Learn More
Syngene™ LED Module for Syngene Gbox Chemi System
Syngene™ LED Module for Syngene Gbox Chemi System for western fluorescence
Brand: Syngene™ LED-GREEN-M x 2
This item is not returnable.
View return policy
Specifications
Green | |
2 | |
LED Module, Multiplex |
Gbox Chemi Systems | |
520nm |