Learn More
Abeba™ Leather Working Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7031/38
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather
- Black
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
Black | |
38 | |
7031 |
Product Content Correction
Product Title
Abeba™ Leather Working Shoes > Size: 38