Learn More
Abeba™ The Original 7050 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7050/47
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance outsole sole
- Sanitized treated
- Leather top layer
- adjustable instep area
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
White | |
47 | |
7050 |
Product Content Correction
Product Title
Abeba™ The Original 7050 Shoes > Size: 47