missing translation for 'onlineSavingsMsg'
Learn More

LCL161, MedChemExpress

MedChemExpress LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.

Encompass_Preferred

1959.66 NOK - 9403.22 NOK

Specifications

Chemical Name or Material LCL161
CAS 1005342-46-0
Color White
Purity Grade Notes Research
Molecular Formula C26H33FN4O3S
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16429167
Encompass_Preferred
View Documents
Medchem Express
HY-15518
5 mg
1959.66 NOK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16426598
Encompass_Preferred
View Documents
Medchem Express
HY-15518
10 mg
2544.40 NOK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16452687
Encompass_Preferred
View Documents
Medchem Express
HY-15518
50 mg
5878.99 NOK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15590593
Encompass_Preferred
View Documents
Medchem Express
HY-15518
100 mg
9403.22 NOK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.
Specifications

Specifications

LCL161
White
C26H33FN4O3S
C[C@H](NC)C(N[C@@H](C1CCCCC1)C(N2[C@H](C3=NC(C(C4=CC=C(F)C=C4)=O)=CS3)CCC2)=O)=O
500.63
Research
Cancer-programmed cell death
1005342-46-0
Research
DMSO : 100 mg/mL (199.75 mM; Need ultrasonic)
500.63
98.65%
Solid
Videos
Safety and Handling

Safety and Handling

H302

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
LCL161, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.