Learn More
LCL161, MedChemExpress
MedChemExpress LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.
Brand: Medchem Express HY-15518
Description
LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.Specifications
LCL161 | |
White | |
C26H33FN4O3S | |
DMSO : 100 mg/mL (199.75 mM; Need ultrasonic) | |
500.63 | |
98.65% | |
Solid |
1005342-46-0 | |
Research | |
10 mg | |
C[C@H](NC)C(N[C@@H](C1CCCCC1)C(N2[C@H](C3=NC(C(C4=CC=C(F)C=C4)=O)=CS3)CCC2)=O)=O | |
500.63 | |
Research | |
Cancer-programmed cell death |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only