missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Sign: Water fog applicator - ISO 7010

Sign compliant to ISO 7010.

39.40 NOK - 104.00 NOK

Specifications

Certifications/Compliance ISO 7010
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
View More Specs

Products 3
Product Code Brand Width (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Height (Metric) Price Quantity & Availability  
15654197
View Documents
Brady™
135955
100 mm 100 mm
39.40 NOK
Each
Estimated Shipment: 13-06-2025
Add to basket
 
15664197
View Documents
Brady™
135956
148 mm 148 mm
104.00 NOK
Each
Estimated Shipment: 13-06-2025
Add to basket
 
15674197
View Documents
Brady™
135957
200 mm 200 mm
54.55 NOK
Each
Estimated Shipment: 13-06-2025
Add to basket
 
Description

Description

  • Laminated polyester
Specifications

Specifications

ISO 7010
Polyester
Permanent Acrylic
-40°C to 110°C
Global
Gloss
1 Ea.
0.099 mm
Square
White on Red
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Laminated Polyester Sign: Water fog applicator - ISO 7010

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.