missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Sign: Water fog applicator - ISO 7010

Sign compliant to ISO 7010.

Brand:  Brady™ 135956

 View more versions of this product

Product Code. 15664197

  • 104.00 NOK / Each
Estimated Shipment: 16-06-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laminated polyester
Specifications

Specifications

ISO 7010
Gloss
1 Ea.
148 mm
148 mm
Square
White on Red
F/F011/NT-SA-148X148/1-B
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Global
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Laminated Polyester Sign: Water fog applicator - ISO 7010 > W x H: 148 x 148 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.