missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase A/LDHA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1815.00 NOK - 4030.00 NOK
Specifications
Antigen | Lactate Dehydrogenase A/LDHA |
---|---|
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329466
|
Bio-Techne
NBP3-16901-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18052884
|
Bio-Techne
NBP3-16901-100UL |
100 μg |
4030.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactate Dehydrogenase A/LDHA Polyclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse samples. It is validated for Western BlotSpecifications
Lactate Dehydrogenase A/LDHA | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience | |
PBS, 50% glycerol, pH7.3 | |
3939 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 | |
A synthetic phosphorylated peptide around Y10 of human Lactate Dehydrogenase A/LDHA (NP_001128711.1). LIYNL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title