missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase A/LDHA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-16901-100UL
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase A/LDHA Polyclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse samples. It is validated for Western BlotSpecifications
Lactate Dehydrogenase A/LDHA | |
Polyclonal | |
Western Blot 1:500 - 1:1000 | |
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 | |
A synthetic phosphorylated peptide around Y10 of human Lactate Dehydrogenase A/LDHA (NP_001128711.1). LIYNL | |
100 μg | |
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience | |
3939 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction