missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT140 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17460-1-AP-150UL
This item is not returnable.
View return policy
Description
IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.Specifications
IFT140 | |
Polyclonal | |
Unconjugated | |
IFT140 | |
c305C8.4, c380F5.1, gs114, IFT140, KIAA0590, WDTC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362124, 106633, 9742 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9PY46, Q96RY7 | |
Ift140 | |
IFT140 Fusion Protein Ag11119 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction