missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Icam-1 Rabbit anti-Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10020-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is raised against mouse Icam-1 which is a cell surface sialoglycoprotein with an apparent molecular weight of 95-100 kDa.
CD54, (MW 90,000) antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.Specifications
Icam-1 | |
Polyclonal | |
Unconjugated | |
Icam1 | |
CD54, ICAM 1, Icam1, Ly 47, MALA 2 | |
Rabbit | |
150 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13597, Q00238 | |
Icam1 | |
Antigen Affinity Chromatography | |
RUO | |
15894, 25464 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction