missing translation for 'onlineSavingsMsg'
Learn More

Icam-1 Rabbit anti-Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4731.57 NOK

Specifications

Antigen Icam-1
Concentration 1.23 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16806173
View Documents
Proteintech
10020-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16896163
View Documents
Proteintech
10020-1-AP-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This antibody is raised against mouse Icam-1 which is a cell surface sialoglycoprotein with an apparent molecular weight of 95-100 kDa.

CD54, (MW 90,000) antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.
Specifications

Specifications

Icam-1
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse
P13597, Q00238
15894, 25464
IgG
Antigen Affinity Chromatography
Antibody
1.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CD54, ICAM 1, Icam1, Ly 47, MALA 2
Icam1
Primary
-20°C
Icam1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.