missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NKp44 ELISA Kit
Human NKp44 quantitates human NKp44 in serum, plasma, supernatant.
5430.00 NOK - 26530.00 NOK
Specifications
Accession Number | CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1 |
---|---|
Assay Range | 13 to 3,000 pg/mL |
Assay Sensitivity | 13 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17105119
|
Invitrogen™
EH340RBX5 |
5 x 96 Tests |
26530.00 NOK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
17104643
|
Invitrogen™
EH340RB |
96 Tests |
5430.00 NOK
Pack of 96 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
Human NKp44 quantitates human NKp44 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NKp44.
CD336 is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.Specifications
CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1 | |
13 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
NCR2 | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
NKp44 | |
4 hr 45 min |
13 to 3,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O95944 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title