Learn More
Invitrogen™ Human NKp44 ELISA Kit
Human NKp44 quantitates human NKp44 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH340RB
 View more versions of this product
Product Code. 17104643
  • 5430.00 NOK / Pack of 96
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Description
Human NKp44 quantitates human NKp44 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NKp44.
CD336 is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1
13 pg/mL
ELISA
Colorimetric Microplate Reader
NCR2
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
13 to 3,000 pg/mL
Biotin
Plasma, Serum, Supernatant
O95944
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL
NKp44
4 hr 45 min