Learn More
R&D Systems Human Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LXSAHM-46
 View more versions of this product
Product Code. 18451260
  • 69120.00 NOK / 1 set
Explore more special offers
Specifications
46 Plex
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Human
GZMB
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems Human Luminex™ Discovery Assay > 46 Plex