Learn More
Korff™ Aluminum Small Rolls
Brand:  Korff™ 60020
Product Code. 10424851
  • 133.00 NOK / 10m
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Specifications
Small Rolls
450mm
Aluminum
Product Title
Korff™ Aluminum Small Rolls > Width: 450mm; Foil Gauge: 0.015mm