missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Desmoglein-2 ELISA Kit

Human Desmoglein-2 quantitates human Desmoglein-2 in serum, plasma, supernatant.

5540.00 NOK - 41590.00 NOK

Specifications

Accession Number AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; Dsg2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Assay Range 16 to 4,000 pg/mL
Assay Sensitivity 16 pg/mL
Conjugate Biotin
Product Type ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17104909
View Documents
Invitrogen™
EH154RBX5
5 x 96 Tests
27070.00 NOK
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
17154433
View Documents
Invitrogen™
EH154RB
96 Tests
5540.00 NOK
Pack of 96
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
 
18577894
View Documents
Invitrogen™
EH154RBX10
10 x 96 Tests
41590.00 NOK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human Desmoglein-2 quantitates human Desmoglein-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Desmoglein-2.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications

Specifications

AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; Dsg2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
16 pg/mL
ELISA
Q14126
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Research Use Only
Desmoglein 2 (DSG2)
4 hr 45 min
16 to 4,000 pg/mL
Biotin
Colorimetric Microplate Reader
DSG2
<10%
HRP
2°C to 8°C
1 hr 20 min
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.