missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Desmoglein-2 ELISA Kit
Human Desmoglein-2 quantitates human Desmoglein-2 in serum, plasma, supernatant.
Brand: Invitrogen™ EH154RBX10
This item is not returnable.
View return policy
Description
Human Desmoglein-2 quantitates human Desmoglein-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Desmoglein-2.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.Specifications
AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; Dsg2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037 | |
16 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
DSG2 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
16 to 4,000 pg/mL | |
Biotin | |
Plasma | |
Q14126 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 µL; Serum, 50 µL; Supernatant, 50 µL | |
Desmoglein 2 (DSG2) | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction