missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPGD Mouse anti-Human, Clone: 3F1F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66798-1-IG-20UL
This item is not returnable.
View return policy
Description
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.Specifications
HPGD | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15428 | |
HPGD | |
HPGD Fusion Protein Ag1499 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
3F1F5 | |
Unconjugated | |
HPGD | |
15 PGDH, HPGD, PGDH, PGDH1, Prostaglandin dehydrogenase 1, SDR36C1 | |
Mouse | |
Protein A | |
RUO | |
3248 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction