missing translation for 'onlineSavingsMsg'
Learn More

HPGD Mouse anti-Human, Clone: 3F1F5, Proteintech

Mouse Monoclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen HPGD
Clone 3F1F5
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16849005
View Documents
Proteintech
66798-1-IG-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16839005
View Documents
Proteintech
66798-1-IG-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.
Specifications

Specifications

HPGD
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
15 PGDH, HPGD, PGDH, PGDH1, Prostaglandin dehydrogenase 1, SDR36C1
HPGD
IgG2b
Protein A
Antibody
3F1F5
Western Blot
Unconjugated
Mouse
Human
P15428
3248
HPGD Fusion Protein Ag1499
Primary
-20°C
HPGD
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.