missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10953-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.Specifications
HMGN2 | |
Polyclonal | |
Unconjugated | |
HMGN2 | |
HMG17, HMGN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3151 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05204 | |
HMGN2 | |
HMGN2 Fusion Protein Ag1401 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction