missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMBS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14797-1-AP-150UL
This item is not returnable.
View return policy
Description
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.Specifications
HMBS | |
Polyclonal | |
Unconjugated | |
HMBS | |
HMBS, hydroxymethylbilane synthase, PBG D, PBGD, Porphobilinogen deaminase, Pre uroporphyrinogen synthase, UPS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15288, 25709, 3145 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08397, P19356, P22907 | |
Hmbs | |
HMBS Fusion Protein Ag6509 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction