Learn More
HLA-DR Mouse, anti-Human, PE, Clone: LN3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60164PE
Encompass_Preferred
 View more versions of this product
Product Code. 17109231
  • 5452.29 NOK / 100 tests
Explore more special offers
Description
Description
  • The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II antigen encoded within the human leukocyte antigen (HLA) complex on chromosome 6
  • HLA-DR is a heterodimeric transmembrane glycoprotein composed of one α (36 kDa) and one β (27 kDa) subunit
  • MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4
  • HLA-DR is primarily expressed on the surface of antigen presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T cells
Specifications
Specifications
HLA-DR
Monoclonal
PE
100 Tests
Rhesus Monkey, Human
B Cells, Dendritic Cells, Macrophages, Monocytes, Other, T Cells
Immunohistochemistry, Flow Cytometry, Functional Assay
LN3
Mouse
Primary
IgG2b κ