missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Mouse, anti-Human, PE, Clone: LN3, STEMCELL Technologies
Mouse Monoclonal Antibody
2449.58 NOK - 5452.29 NOK
Specifications
Antigen | HLA-DR |
---|---|
Clone | LN3 |
Applications | Immunohistochemistry, Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17119231
|
Stemcell Technologies
60164PE.1 |
25 Tests |
2449.58 NOK
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17109231
|
Stemcell Technologies
60164PE |
100 Tests |
5452.29 NOK
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II antigen encoded within the human leukocyte antigen (HLA) complex on chromosome 6
- HLA-DR is a heterodimeric transmembrane glycoprotein composed of one α (36 kDa) and one β (27 kDa) subunit
- MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4
- HLA-DR is primarily expressed on the surface of antigen presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T cells
Specifications
HLA-DR | |
Immunohistochemistry, Flow Cytometry, Functional Assay | |
PE | |
Rhesus Monkey, Human | |
Primary |
LN3 | |
Monoclonal | |
Mouse | |
IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title