Learn More
HLA-DR Mouse, anti-Human, PE, Clone: LN3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60164PE.1
This item is not returnable.
View return policy
Description
- The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II antigen encoded within the human leukocyte antigen (HLA) complex on chromosome 6
- HLA-DR is a heterodimeric transmembrane glycoprotein composed of one α (36 kDa) and one β (27 kDa) subunit
- MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4
- HLA-DR is primarily expressed on the surface of antigen presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T cells
Specifications
HLA-DR | |
Monoclonal | |
PE | |
25 Tests | |
Rhesus Monkey, Human | |
B Cells, Dendritic Cells, Macrophages, Monocytes, Other, T Cells |
Immunohistochemistry, Flow Cytometry, Functional Assay | |
LN3 | |
Mouse | |
Primary | |
IgG2b κ |