missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Buffer, Powder, Cell Culture Grade, MP Biomedicals
Powdered Storage temperature: 15–30°C
Brand: MP Biomedicals 091588413
Quantity | 20 g |
---|
This item is not returnable.
View return policy
Description

Specifications
7365-45-9 | |
C8H18N2O4S | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
23831 | |
Cell Culture |
20 g | |
MFCD00006158 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
HEPES Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HEPES Buffer, Powder, Cell Culture Grade, MP Biomedicals > 20g
Spot an opportunity for improvement?Share a Content Correction