missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Gypsy Moth Diet Without Agar
Previously been known as the high wheat germ diet.
Brand: MP Biomedicals 0296029404
This item is not returnable.
View return policy
Description
- Available with agar included, packaged separately or omitted completely
- Methyl paraben and sorbic acid added as preservatives

Specifications
Not Autoclavable | |
Single-use | |
Non-sterile | |
9 in. |
Animal Diet | |
4 L | |
Room Temperature | |
0.2286 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals Gypsy Moth Diet Without Agar > Gypsy Moth Diet Without Agar, 4 L
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction