missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Gypsy Moth Diet Without Agar
Previously been known as the high wheat germ diet.
321.00 NOK - 974.00 NOK
Specifications
Product Type | Animal Diet |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Sterility | Non-sterile |
Storage Requirements | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447231
|
MP Biomedicals
0296029401 |
1 L |
321.00 NOK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480056
|
MP Biomedicals
0296029404 |
4 L |
974.00 NOK
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Available with agar included, packaged separately or omitted completely
- Methyl paraben and sorbic acid added as preservatives
Specifications
Animal Diet | |
Single-use | |
Room Temperature | |
0.2286 m |
Not Autoclavable | |
Non-sterile | |
9 in. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title