Learn More
Granzyme K Mouse anti-Human, Unconjugated, Clone: 1C3A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67272-1-IG-20UL
This item is not returnable.
View return policy
Description
The Granzyme K antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Granzyme K. This antibody recognizes human antigen. The Granzyme K antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Granzyme K | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human |
Immunohistochemistry, Immunofluorescence, ELISA | |
1C3A4 | |
Unconjugated | |
20 μL | |
3003 | |
IgG2b |