missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme K Mouse anti-Human, Unconjugated, Clone: 1C3A4, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | Granzyme K |
---|---|
Clone | 1C3A4 |
Concentration | 500 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849805
|
Proteintech
67272-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839805
|
Proteintech
67272-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Granzyme K antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Granzyme K. This antibody recognizes human antigen. The Granzyme K antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Granzyme K | |
500 μg/mL | |
Monoclonal | |
Mouse | |
3003 | |
Primary |
1C3A4 | |
Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title