missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15752-1-AP-150UL
This item is not returnable.
View return policy
Description
Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII). May act at an RNAP assembly step prior to nuclear import.Specifications
GPN1 | |
Polyclonal | |
Unconjugated | |
GPN1 | |
ATPBD1A, GPN loop GTPase 1, GPN1, MBD2 interacting protein, MBDIN, NTPBP, XAB1, XPA binding protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11321 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HCN4 | |
GPN1 | |
GPN1 Fusion Protein Ag8252 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction