missing translation for 'onlineSavingsMsg'
Learn More

GPN1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen GPN1
Concentration 0.23 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16855074
View Documents
Proteintech
15752-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16845074
View Documents
Proteintech
15752-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII). May act at an RNAP assembly step prior to nuclear import.
Specifications

Specifications

GPN1
Western Blot
Unconjugated
Rabbit
Human
Q9HCN4
11321
GPN1 Fusion Protein Ag8252
Primary
-20°C
GPN1
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ATPBD1A, GPN loop GTPase 1, GPN1, MBD2 interacting protein, MBDIN, NTPBP, XAB1, XPA binding protein 1
GPN1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.