missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reduced, Cell Culture Reagent, ≥98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
Description
- Glutathione suppresses human immunodeficiency virus expression in chronically infected monocytic cells.
- It is a useful tripeptide involved in many aspects of metabolism, including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds.
- Glutathione is an endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst.
- Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center.
- Glutathione also forms disulfide bonds with cysteine residues in proteins.
- Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.
- Typical Working Concentration: May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.

Specifications
Specifications
Melting Point | 195°C |
Color | White |
Assay Percent Range | >98% |
Quantity | 10 g |
Solubility Information | 292.5 mg/mL |
Formula Weight | 307.32 |
Percent Purity | ≥98% |
Grade | Cell Culture |
Physical Form | Powder |
Chemical Name or Material | Glutathione Reduced, Cell Culture Reagent |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glutathione Reduced, Cell Culture Reagent, ≥98%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction