missing translation for 'onlineSavingsMsg'
Learn More
Glutathione Reduced Crystalline MP Biomedicals Product Code.: 11304269

Glutathione Reduced Crystalline MP Biomedicals

Product Code. 11304269 Shop All MP Biomedicals Products
50 g, 50g
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
1g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11304269

Brand: MP Biomedicals 0210181450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Color White
Assay Percent Range >98%
Quantity 50 g
Formula Weight 307.3
Physical Form Powder
Chemical Name or Material Glutathione, Reduced
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glutathione Reduced Crystalline MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.