Learn More
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted
Pycnometers acc. to Gay Lussac, unadjusted
Brand:  Lenz Laborglasintrumente™ 05630028
 View more versions of this product
Product Code. 10000281
  • 400.00 NOK / Each
Estimated Shipment: 05-05-2025
to see stock.
Explore more special offers
Description
  • Gay Lussac pycnometers acc. to DIN ISO 3507 and in addition to standard
  • Unadjusted
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Specifications
1
50 mL
Borosilicate Glass (3.3)
Product Title
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted > Capacity (Metric): 50 cm³