Learn More
BRAND™ Gay-Lussac Calibrated Density Bottle
Borosilicate glass construction. The volume in cm3 is specified to a precision of 3 decimal places.
Brand:  BRAND™ 43338
987.38 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10221981
  • 1260.00 NOK / Each
Only null left
Estimated Shipment: 04-08-2025
Description
  • Borosilicate glass 3.3.
  • DIN ISO 3507
  • Gay-Lussac style
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • The volume in cmÂł is specified to a precision of 3 decimal places
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
100 mL
Borosilicate Glass (3.3)
Gay-Lussac Density Bottle
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Gay-Lussac Calibrated Density Bottle > Volume: 100mL