missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | GABRA5 |
---|---|
Concentration | 270 μg/mL |
Applications | Immunohistochemistry, Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17741079
|
Proteintech
27929-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17731079
|
Proteintech
27929-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABRA5 Polyclonal antibody specifically detects GABRA5 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISASpecifications
GABRA5 | |
Immunohistochemistry, Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
IgG | |
Antigen affinity purification | |
Antibody |
270 μg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse, Rat | |
2558 | |
Primary | |
Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title