missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27929-1-AP-150UL
This item is not returnable.
View return policy
Description
GABRA5 Polyclonal antibody specifically detects GABRA5 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISASpecifications
GABRA5 | |
Polyclonal | |
Unconjugated | |
BC146493 | |
Antigen affinity purification | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot, ELISA | |
270 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Rabbit | |
150 μL | |
2558 | |
Store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction