Learn More
FT011, MedChemExpress

MedChemExpress FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.

POA - 7348.74 NOK
Specifications
Chemical Name or Material | FT011 |
---|---|
CAS | 1001288-58-9 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C20H17NO5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490016
![]() |
Medchem Express
HY-100495 |
2 mg | |||||||
16474346
![]() |
Medchem Express
HY-100495 |
100 mg |
7348.74 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16443048
![]() |
Medchem Express
HY-100495 |
10 mg |
1311.71 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415787
![]() |
Medchem Express
HY-100495 |
5 mg |
821.80 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16409616
![]() |
Medchem Express
HY-100495 |
50 mg |
4567.28 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.Specifications
FT011 | |
Light Yellow | |
C20H17NO5 | |
O=C(O)C1=CC=CC=C1NC(/C=C/C2=CC=C(OCC#C)C(OC)=C2)=O | |
351.35 | |
Research | |
COVID-19-immunoregulation |
1001288-58-9 | |
Research | |
DMSO : 100 mg/mL (284.62 mM; Need ultrasonic) | |
351.35 | |
99.63% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.