Learn More
FT011, MedChemExpress
MedChemExpress FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.
Brand: Medchem Express HY-100495
Description
FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.Specifications
FT011 | |
Light Yellow | |
C20H17NO5 | |
DMSO : 100 mg/mL (284.62 mM; Need ultrasonic) | |
351.35 | |
99.63% | |
Solid |
1001288-58-9 | |
Research | |
50 mg | |
O=C(O)C1=CC=CC=C1NC(/C=C/C2=CC=C(OCC#C)C(OC)=C2)=O | |
351.35 | |
Research | |
COVID-19-immunoregulation |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only