Learn More
Wesbart™ Front Access Freezer Racks
Wesbart Front Access Freezer Racks are used to store boxes.
Brand: Wesbart™ AH179/2UT
This item is not returnable.
View return policy
Description
- ULT freezer racks
- For upright freezers to store boxes
Specifications
2 Trays | |
Aluminum Front Access Rack | |
3 | |
Yes | |
Store Loose Items | |
560 x 140 x 415 mm |
Freezer Rack | |
Autoclavable | |
Rectangular | |
Non-sterile | |
Freezers |