missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD3 Monoclonal Antibody (5G9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515731
This item is not returnable.
View return policy
Description
MA5-15731 targets FOXD3 in indirect ELISA, IHC and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15731 immunogen is purified recombinant fragment of human FOXD3 expressed in E. Coli. MA5-15731 detects FOXD3 which has a predicted molecular weight of approximately 50kDa.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
Specifications
FOXD3 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q61060, Q9UJU5 | |
FOXD3 | |
Purified recombinant fragment of human FOXD3 expressed in E. Coli. | |
RUO | |
15221, 27022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
5G9 | |
Unconjugated | |
FOXD3 | |
AIS1; CWH3; forkhead box D3; forkhead box protein D3; FOXD3; Genesis; hepatocyte nuclear factor 3 forkhead homolog 2; HFH2; HFH-2; HNF3/FH transcription factor genesis; HNF-3/forkhead homolog 2; VAMAS2 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXD3 Monoclonal Antibody (5G9), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction