Learn More
FOXP3 Monoclonal Antibody (3G3), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA141628
Description
MA1-41628 is derived from full-length His-tagged mouse FOXP3. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein.
FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications
FOXP3 | |
Monoclonal | |
1.85 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
Q99JB6, Q9BZS1 | |
Foxp3 | |
Full-length, His-tagged murine FOXP3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
3G3 | |
FITC | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Mouse | |
Protein G | |
RUO | |
20371, 50943 | |
4° C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.