missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FOSL2 (Human) Recombinant Protein
Human FOSL2 full-length ORF ( AAH08899, 1 a.a. - 122 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002355-P01.10ug
This item is not returnable.
View return policy
Description
- Sequence: MSVGLDLPQMLPGSPSPSLKEAFAEDGEAGEGGGRPSLEWRLQQLLPQHPLSLSWLLTSPQGTGPFLPSVVICHLLDQVLSLLHSPVPTPVHSSGPGSKQAVNSWPELSLWLVAHAPFLVVC
Specifications
AAH08899 | |
FOS-like antigen 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FOSL2 | |
GST |
2355 | |
Wheat germ expression system | |
10 μg | |
FLJ23306, FRA2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |