missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Flask Platforms for use with ES-20/80

The Grant™ interchangeable flask platforms are for use with the ES-20/80 Orbital Shaker-Incubator. The platforms have clamps to securely hold the flasks in position during vigorous mixing.

Brand:  Grant Instruments™ HSP-9/500

 View more versions of this product

Product Code. 17775019

  • 6130.00 NOK / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Interchangeable
  • Clamps securely hold flasks in place during vigorous mixing
  • Optional accessories for the ES-20/80 Orbital Shaker-Incubator
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9 x 500 mL Flasks
7.3 lb.
Platform for 500 mL Flasks (9 x 500 mL Flasks. ES-20/80 and ES-20/80C)
36 x 40 x 10 cm
3.3 kg
ES-20/80 and ES-20/80C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Grant Instruments™ Flask Platforms for use with ES-20/80 > Holds: 9 x 500 mL Flasks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.