Learn More
BRAND™ Filling Valve with Seal
Includes sealing washer. For use with Dispensette™ III, seripettor™ pro and Digital Burette.
Brand: BRAND™ 6697
208.24 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Dispensette™ III, seripettor™ pro and Digital Burette
- Volume range: 0.5, 1, 2, 5, 10mL
Specifications
For Burettes | |
1 Each | |
With Sealing Washer | |
Filling Valve |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Filling Valve with Seal > Capacity: 0.5, 1, 2, 5, 10 mL, with Seal