Learn More
Brand™ Rubber Bellows for Automatic Burettes
Shop All Brand Products
Click to view available options
Color:
Orange
Unit Size:
Each
Description
- Orange colored
- Single bulb
- Provided individually
Specifications
Specifications
Color | Orange |
For Use With (Equipment) | Automatic Burette |
Material | Rubber |
No. per Pack | 1 |
Includes | Tubing, Valve |
Type | Bellows |
Product Content Correction
Product Title
Brand™ Rubber Bellows for Automatic Burettes >